siRNA Details
| Gene Symbol: Snx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207213.2 | NP_997096.2 | 4 | 515 | ||
| GenBank mRNA | AY601646.1 | - | 3 | 734 | ||
| BC057063.1 | - | 3 | 263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx25| Gene Symbol: Snx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207213.2 | NP_997096.2 | 4 | 515 | ||
| GenBank mRNA | AY601646.1 | - | 3 | 734 | ||
| BC057063.1 | - | 3 | 263 | |||