siRNA Details
| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081240.3 | NP_001074709.1 | 7 | 1125 | ||
| GenBank mRNA | AK045315.1 | - | 7 | 1115 | ||
| AK154546.1 | - | 7 | 1069 | |||
| AK161023.1 | - | 7 | 1034 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt10| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081240.3 | NP_001074709.1 | 7 | 1125 | ||
| GenBank mRNA | AK045315.1 | - | 7 | 1115 | ||
| AK154546.1 | - | 7 | 1069 | |||
| AK161023.1 | - | 7 | 1034 | |||