siRNA Details
| Gene Symbol: Ankrd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281974.1 | NP_001268903.1 | 7 | 1159 | ||
| NM_001281975.1 | NP_001268904.1 | 7 | 1209 | |||
| NM_133971.3 | NP_598732.2 | 6 | 1047 | |||
| GenBank mRNA | AK081754.1 | - | 7 | 1207 | ||
| AK082875.1 | - | 6 | 1044 | |||
| AK084456.1 | - | 7 | 1157 | |||
| AK086012.1 | - | 4 | 1403 | |||
| AK088371.1 | - | 6 | 997 | |||
| BC002198.1 | - | 6 | 979 | 3489133 | ||

Ankrd10
Genomic Map