siRNA Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271873.1 | NP_001258802.1 | 4 | 513 | ||
| NM_133979.3 | NP_598740.1 | 4 | 513 | |||
| GenBank mRNA | AK043011.1 | - | 4 | 476 | ||
| AK046091.1 | - | 4 | 465 | |||
| AK079622.1 | - | 4 | 464 | |||
| AK080947.1 | - | 4 | 465 | |||
| AK138414.1 | - | 4 | 499 | |||
| AK195077.1 | - | 2 | 88 | |||
| BC099688.1 | - | 4 | 436 | 6313494 | ||

Ano10
Genomic Map