siRNA Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271873.1 | NP_001258802.1 | 9 | 1339 | ||
| NM_133979.3 | NP_598740.1 | 9 | 1513 | |||
| GenBank mRNA | AK028391.1 | - | 4 | 458 | ||
| AK043011.1 | - | 9 | 1477 | |||
| AK046091.1 | - | 9 | 1465 | |||
| AK054163.1 | - | 5 | 803 | |||
| AK079622.1 | - | 9 | 1464 | |||
| AK080947.1 | - | 9 | 1465 | |||
| AK138414.1 | - | 9 | 1588 | |||
| BC026421.1 | - | 4 | 553 | |||
| BC099688.1 | - | 9 | 1262 | 6313494 | ||

Ano10
Genomic Map