siRNA Details
| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC021449.1 | - | 4 | 820 | 5044564 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a13| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC021449.1 | - | 4 | 820 | 5044564 | |