siRNA Details
| Gene Symbol: OAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032731.1 | NP_001027903.1 | 2 | 338 | ||
| NM_002535.2 | NP_002526.2 | 2 | 338 | |||
| NM_016817.2 | NP_058197.2 | 2 | 338 | |||
| GenBank mRNA | AK292796.1 | - | 2 | 227 | ||
| AK292906.1 | - | 2 | 255 | |||
| BC010625.1 | - | 2 | 324 | |||
| BC023637.2 | - | 2 | 259 | |||
| BC049215.1 | - | 2 | 239 | 5517804 | ||
| BC053870.1 | - | 2 | 261 | |||
| BC069052.1 | - | 2 | 250 | |||
| BE885222.1 | - | 395 | ||||
| M87284.1 | - | 2 | 217 | |||
| M87434.1 | - | 2 | 217 | |||

OAS2
Genomic Map