siRNA Details
| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028874.2 | NP_083150.1 | 2 | 2013 | ||
| GenBank mRNA | AK044307.1 | - | 2 | 101 | ||
| AK122231.1 | - | 2 | 2234 | |||
| BC004635.1 | - | 2 | 485 | |||
| BC051164.1 | - | 2 | 2233 | |||
| BC063262.1 | - | 2 | 2013 | 6405897 | ||

Snx19
Genomic Map