siRNA Details
| Gene Symbol: Cd276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133983.4 | NP_598744.1 | 2 | 577 | ||
| GenBank mRNA | AK031354.1 | - | 2 | 308 | ||
| AK155114.1 | - | 3 | 663 | |||
| AY190318.1 | - | 2 | 392 | |||
| BC019436.1 | - | 2 | 481 | 3674228 | ||
| BC056608.1 | - | 2 | 578 | 6397680 | ||
| DQ832276.1 | - | 3 | 676 | |||

Cd276
Genomic Map