siRNA Details
| Gene Symbol: Bbs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175325.3 | NP_780534.1 | 8 | 581 | ||
| GenBank mRNA | AK039560.1 | - | 8 | 530 | ||
| AK089970.1 | - | 8 | 536 | |||
| AK134734.1 | - | 8 | 534 | |||
| BC055797.1 | - | 8 | 501 | |||
| BC089507.1 | - | 8 | 515 | |||
| BC092531.1 | - | 8 | 525 | |||
| BC145771.1 | - | 8 | 543 | 40131074 | ||
| BC145773.1 | - | 8 | 543 | 40131076 | ||

Bbs4
Genomic Map