siRNA Details
| Gene Symbol: Bbs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175325.3 | NP_780534.1 | 10 | 760 | ||
| GenBank mRNA | AK039560.1 | - | 10 | 709 | ||
| AK089970.1 | - | 10 | 715 | |||
| AK134734.1 | - | 10 | 713 | |||
| BC055797.1 | - | 10 | 680 | |||
| BC089507.1 | - | 10 | 694 | |||
| BC092531.1 | - | 10 | 704 | |||
| BC145771.1 | - | 10 | 722 | 40131074 | ||
| BC145773.1 | - | 10 | 722 | 40131076 | ||

Bbs4
Genomic Map