siRNA Details
Gene Symbol: B630019K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046767.1 | - | 1 | 1292 | ||
BC055780.1 | - | 1 | 1213 | 6403722 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B630019K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046767.1 | - | 1 | 1292 | ||
BC055780.1 | - | 1 | 1213 | 6403722 |