siRNA Details
| Gene Symbol: Sept10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024910.3 | NP_001020081.2 | 9 | 1694 | ||
| NM_001024911.2 | NP_001020082.1 | 9 | 1694 | |||
| GenBank mRNA | AK076569.1 | - | 9 | 1695 | ||
| BC053752.1 | - | 9 | 1308 | 6506619 | ||

Sept10
Genomic Map