siRNA Details
| Gene Symbol: Rdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153133.2 | NP_694773.1 | 1 | 154 | ||
| GenBank mRNA | AB081322.1 | - | 1 | 92 | ||
| AF372838.1 | - | 1 | 154 | |||
| BC056642.1 | - | 1 | 164 | |||
| BC141318.1 | - | 1 | 134 | 9055930 | ||

Rdh9
Genomic Map