siRNA Details
| Gene Symbol: Upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133995.4 | NP_598756.1 | 4 | 546 | ||
| GenBank mRNA | AK149373.1 | - | 4 | 443 | ||
| AK149480.1 | - | 4 | 443 | |||
| AK165429.1 | - | 4 | 477 | |||
| BC021388.1 | - | 4 | 431 | 5055165 | ||
| BC024447.1 | - | 2 | 97 | |||

Upb1
Genomic Map