siRNA Details
| Gene Symbol: Fig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133999.1 | NP_598760.1 | 2 | 283 | ||
| GenBank mRNA | AK041196.1 | - | 2 | 284 | ||
| AK155392.1 | - | 2 | 280 | |||
| AK169881.1 | - | 2 | 247 | |||
| AK172130.1 | - | 2 | 277 | |||
| AK172341.1 | - | 2 | 247 | |||
| AK194775.1 | - | 2 | 83 | |||
| BC015295.1 | - | 2 | 283 | 4221630 | ||
| BC031887.1 | - | 2 | 282 | 5363304 | ||

Fig4
Genomic Map