siRNA Details
| Gene Symbol: Ttc41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003910.2 | NP_001003910.2 | 2 | 480 | ||
| NM_001003939.2 | NP_001003939.2 | 3 | 589 | |||
| NM_153595.3 | NP_705823.3 | 1 | 273 | |||
| GenBank mRNA | AK133195.1 | - | 2 | 420 | ||
| AY651019.1 | - | 4 | 672 | |||
| AY651020.1 | - | 1 | 120 | |||
| AY651021.1 | - | 1 | 120 | |||
| BC030307.1 | - | 3 | 567 | |||

Ttc41
Genomic Map