siRNA Details
Gene Symbol: Ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134009.3 | NP_598770.1 | 8 | 1187 | ||
GenBank mRNA | AK019409.1 | - | 8 | 1057 | ||
AK048954.1 | - | 8 | 1181 | |||
AK128887.1 | - | 8 | 1188 | |||
BC019501.1 | - | 8 | 1187 | 4193809 |
