siRNA Details
| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134009.3 | NP_598770.1 | 2, 3 | 482 | ||
| GenBank mRNA | AK019409.1 | - | 2, 3 | 352 | ||
| AK048954.1 | - | 2, 3 | 476 | |||
| AK128887.1 | - | 2, 3 | 483 | |||
| BC019501.1 | - | 2, 3 | 482 | 4193809 | ||

Ncln
Genomic Map