siRNA Details
| Gene Symbol: Fbxw11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271347.1 | NP_001258276.1 | 3 | 653 | ||
| NM_001271348.1 | NP_001258277.1 | 3 | 614 | |||
| NM_001271349.1 | NP_001258278.1 | 2 | 551 | |||
| NM_134015.3 | NP_598776.1 | 4 | 716 | |||
| GenBank mRNA | AB093260.1 | - | 2 | 358 | ||
| AK041532.1 | - | 3 | 446 | |||
| AK149139.1 | - | 3 | 436 | |||
| AK152181.1 | - | 3 | 376 | |||
| AK160086.1 | - | 2 | 352 | |||
| AK166226.1 | - | 3 | 311 | |||
| AK168667.1 | - | 2 | 368 | |||
| AK218380.1 | - | 1 | 41 | |||
| AY038079.1 | - | 4 | 380 | |||
| BC034261.1 | - | 4 | 396 | 4237375 | ||
| CA326854.1 | - | 4 | 716 | |||
| CB526764.1 | - | 4 | 716 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106993, XM_006246092, XM_006246093, XM_006246094.

Fbxw11
Genomic Map