siRNA Details
| Gene Symbol: Smg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002764.1 | NP_001002764.1 | 5, 6 | 2300 | ||
| GenBank mRNA | AK129203.1 | - | 4, 5 | 919 | ||
| BC037179.1 | - | 4, 5 | 652 | |||
| BC047279.1 | - | 5, 6 | 2312 | |||
| BC066040.1 | - | 5, 6 | 2300 | 6410245 | ||
| BC077715.1 | - | 5, 6 | 2263 | |||

Smg6
Genomic Map