siRNA Details
| Gene Symbol: Slc35e4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153142.3 | NP_694782.1 | 2 | 2436 | ||
| GenBank mRNA | AK039786.1 | - | 2 | 2437 | ||
| AK218784.1 | - | 1 | 62 | |||
| BC020181.1 | - | 2 | 1470 | 4488513 | ||
| BC095957.1 | - | 2 | 344 | |||

Slc35e4
Genomic Map