siRNA Details
| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134021.2 | NP_598782.1 | 5 | 619 | ||
| GenBank mRNA | AK154004.1 | - | 5 | 559 | ||
| AK154443.1 | - | 5 | 380 | |||
| AK164667.1 | - | 5 | 620 | |||
| BC010785.1 | - | 5 | 558 | 4210325 | ||
| BC026564.1 | - | 5 | 586 | 4971745 | ||
| CK625276.1 | - | 5 | 428 | |||

Pnpo
Genomic Map