siRNA Details
| Gene Symbol: Tbc1d10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134023.1 | NP_598784.1 | 4 | 545 | ||
| GenBank mRNA | AK140872.1 | - | 4 | 557 | ||
| AK153677.1 | - | 527 | ||||
| AK220243.1 | - | 556 | ||||
| BC018300.1 | - | 4 | 545 | 4193046 | ||
| BC029773.1 | - | 2 | 109 | |||
| BC037230.1 | - | 4 | 528 | 2650923 | ||

Tbc1d10a
Genomic Map