siRNA Details
| Gene Symbol: Pex12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134025.3 | NP_598786.1 | 3 | 787 | ||
| GenBank mRNA | AK035552.1 | - | 3 | 788 | ||
| AK041175.1 | - | 3 | 883 | |||
| AK153798.1 | - | 2 | 994 | |||
| AK166413.1 | - | 3 | 831 | |||
| BC021800.1 | - | 2 | 611 | 3671142 | ||

Pex12
Genomic Map