siRNA Details
| Gene Symbol: Mien1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025559.2 | NP_079835.1 | 4 | 671 | ||
| GenBank mRNA | AK002959.1 | - | 4 | 654 | ||
| AK007795.1 | - | 4 | 654 | |||
| AK009922.1 | - | 4 | 649 | |||
| AK041314.1 | - | 4 | 653 | |||
| BC021589.1 | - | 4 | 655 | 5100542 | ||
| CA530220.1 | - | 3 | 497 | |||

Mien1
Genomic Map