siRNA Details
| Gene Symbol: Cuedc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172099.1 | NP_001165570.1 | 7 | 1289 | ||
| NM_198013.3 | NP_932130.2 | 8 | 1487 | |||
| GenBank mRNA | AK049250.1 | - | 8 | 1471 | ||
| BC024476.1 | - | 8 | 1336 | 4976954 | ||
| BC024529.1 | - | 4 | 288 | |||
| BC029708.1 | - | 8 | 1420 | 5041450 | ||
| BC131989.1 | - | 7 | 1260 | 40130266 | ||
| BC131991.1 | - | 7 | 1260 | 40130268 | ||
| BC145206.1 | - | 7 | 1260 | 9053757 | ||
| CF950088.1 | - | 4 | 349 | |||

Cuedc1
Genomic Map