siRNA Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029636.3 | NP_083912.2 | 3 | 264 | ||
| GenBank mRNA | AF456461.1 | - | 3 | 295 | ||
| AY014776.1 | - | 3 | 264 | |||
| BC099415.1 | - | 3 | 271 | 30786803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsq| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029636.3 | NP_083912.2 | 3 | 264 | ||
| GenBank mRNA | AF456461.1 | - | 3 | 295 | ||
| AY014776.1 | - | 3 | 264 | |||
| BC099415.1 | - | 3 | 271 | 30786803 | ||