siRNA Details
| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045515.1 | NP_001038980.1 | 33 | 4616 | ||
| NM_001164483.1 | NP_001157955.1 | 32 | 4685 | |||
| GenBank mRNA | AB093272.1 | - | 32 | 4502 | ||
| CJ048086.1 | - | 1 | 124 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU067566.1 | - | 641 | Chr. - - [-] on Build | ||

Synj1
Genomic Map