siRNA Details
| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138595.2 | NP_613061.1 | 14 | 1874 | ||
| GenBank mRNA | AK080789.1 | - | 14 | 1846 | ||
| AK080937.1 | - | 14 | 1863 | |||
| AK139929.1 | - | 14 | 1866 | |||
| BC017135.1 | - | 14 | 1851 | 4511573 | ||

Gldc
Genomic Map