siRNA Details
| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172549.3 | NP_766137.2 | 4 | 576 | ||
| GenBank mRNA | AK168811.1 | - | 4 | 536 | ||
| BC039933.1 | - | 4 | 403 | |||
| BC057551.1 | - | 4 | 504 | |||
| BC150825.1 | - | 3 | 140 | 9087736 | ||
| CN722898.1 | - | 4 | 576 | |||

Cabin1
Genomic Map