siRNA Details
| Gene Symbol: Kdm3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038695.3 | NP_001033784.2 | 6 | 670 | ||
| NM_173001.3 | NP_766589.1 | 5 | 697 | |||
| GenBank mRNA | AK082379.1 | - | 1 | 251 | ||
| AK129204.1 | - | 6 | 658 | |||
| AK144825.1 | - | 5 | 698 | |||
| BC032256.1 | - | 6 | 646 | |||
| BC059264.1 | - | 5 | 580 | 6409698 | ||
| DQ323991.1 | - | 2 | 135 | |||

Kdm3a
Genomic Map