siRNA Details
| Gene Symbol: Zfp120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023266.4 | NP_075755.4 | 3 | 281 | ||
| NM_181266.2 | NP_851783.2 | 4 | 345 | |||
| GenBank mRNA | AF211867.1 | - | 4 | 345 | ||
| AF211868.1 | - | 4 | 429 | |||
| AF211869.1 | - | 3 | 281 | |||
| AF242378.1 | - | 3 | 266 | |||
| AK033399.1 | - | 4 | 342 | |||
| AK132318.1 | - | 4 | 351 | |||
| BC013500.1 | - | 4 | 348 | |||
| BC141128.1 | - | 4 | 335 | 9055740 | ||

Zfp120
Genomic Map