siRNA Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027088.2 | NP_081364.1 | 4 | 313 | ||
| GenBank mRNA | AB047820.1 | - | 4 | 214 | ||
| AK079374.1 | - | 4 | 329 | |||
| AK129108.1 | - | 4 | 221 | |||
| AK170576.1 | - | 4 | 314 | |||
| AK171561.1 | - | 4 | 315 | |||
| BC042489.1 | - | 4 | 264 | |||
| BC050901.1 | - | 4 | 276 | 6402443 | ||

Bap1
Genomic Map