siRNA Details
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005014.2 | NP_005005.1 | 3 | 1556 | ||
| GenBank mRNA | AB000114.1 | - | 3 | 1287 | ||
| AK225922.1 | - | 3 | 1401 | |||
| AK313055.1 | - | 3 | 1395 | |||
| AY358872.1 | - | 3 | 1454 | |||
| BC046356.1 | - | 3 | 1458 | 5239983 | ||

OMD
Genomic Map