siRNA Details
| Gene Symbol: Smek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134034.2 | NP_598795.2 | 8 | 1721 | ||
| GenBank mRNA | AK037253.1 | - | 8 | 1653 | ||
| AK041513.1 | - | 8 | 1666 | |||
| AK044742.1 | - | 8 | 1664 | |||
| AK090014.1 | - | 8 | 1697 | |||
| AK129344.1 | - | 8 | 1632 | |||
| AK132660.1 | - | 8 | 1243 | |||
| AK165940.1 | - | 8 | 1646 | |||
| AK166627.1 | - | 8 | 1648 | |||
| BC006870.1 | - | 8 | 1632 | 3595439 | ||

Smek2
Genomic Map