siRNA Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170671.2 | NP_733771.2 | 13 | 1785 | ||
| GenBank mRNA | AK029929.1 | - | 12 | 1961 | ||
| AK166780.1 | - | 6 | 933 | |||
| BC027327.1 | - | 9 | 1146 | 3991131 | ||
| BC050808.1 | - | 2 | 308 | |||

Mycbpap
Genomic Map