siRNA Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178911.4 | NP_849242.1 | 11 | 1568 | ||
| GenBank mRNA | AK079549.1 | - | 11 | 1527 | ||
| AK079878.1 | - | 11 | 1570 | |||
| AK151551.1 | - | 11 | 1802 | |||
| AK152969.1 | - | 10 | 1417 | |||
| AK170362.1 | - | 11 | 1525 | |||
| BC058565.1 | - | 11 | 1518 | 6442038 | ||

Pld4
Genomic Map