siRNA Details
| Gene Symbol: Jkamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205067.1 | NP_001191996.1 | 2 | 375 | ||
| NM_024205.2 | NP_077167.1 | 2 | 393 | |||
| GenBank mRNA | AK004555.1 | - | 2 | 330 | ||
| BC002238.1 | - | 2 | 109 | 3491879 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106738.
Set Membership: |
|

Jkamp
Genomic Map