siRNA Details
| Gene Symbol: Tecpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081057.2 | NP_001074526.1 | 14 | 3191 | ||
| NM_001289510.1 | NP_001276439.1 | 13 | 3139 | |||
| NM_175336.3 | NP_780545.2 | 14 | 3301 | |||
| GenBank mRNA | AK129113.1 | - | 13 | 3143 | ||
| AK147592.1 | - | 13 | 3139 | |||
| BC041778.1 | - | 11 | 2783 | |||
| BC138354.1 | - | 13 | 3134 | 8861374 | ||
| BC145250.1 | - | 13 | 3244 | |||

Tecpr2
Genomic Map