siRNA Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289572.1 | NP_001276501.1 | 11 | 1547 | ||
| NM_001289573.1 | NP_001276502.1 | 10 | 1142 | |||
| NM_001289574.1 | NP_001276503.1 | 12 | 1709 | |||
| NM_178914.4 | NP_849245.2 | 12 | 1643 | |||
| GenBank mRNA | AK036359.1 | - | 12 | 1627 | ||
| AK077500.1 | - | 12 | 1554 | |||
| AK153698.1 | - | 10 | 1118 | |||
| AK161308.1 | - | 11 | 1493 | |||
| BC046960.1 | - | 12 | 1601 | 6391450 | ||
| DQ196101.1 | - | 11 | 1373 | |||

Spata7
Genomic Map