siRNA Details
| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028821.3 | NP_083097.2 | 5 | 294 | ||
| GenBank mRNA | AK005826.1 | - | 5 | 380 | ||
| BC070454.1 | - | 251 | ||||
| BC125392.1 | - | 4 | 276 | 40129907 | ||
| BC125394.1 | - | 4 | 272 | 40129909 | ||
| BY126068.1 | - | 2 | 90 | |||

Dnal1
Genomic Map