siRNA Details
| Gene Symbol: Rnf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146025.1 | NP_001139497.1 | 9 | 1454 | ||
| NM_001146026.1 | NP_001139498.1 | 9 | 1379 | |||
| NM_001146027.1 | NP_001139499.1 | 9 | 1245 | |||
| NM_134064.2 | NP_598825.2 | 9 | 1387 | |||
| GenBank mRNA | AK147349.1 | - | 9 | 1380 | ||
| AK160090.1 | - | 9 | 1238 | |||
| AK160115.1 | - | 9 | 1238 | |||
| AK161651.1 | - | 9 | 1369 | |||
| AK171943.1 | - | 9 | 1236 | |||
| AK172257.1 | - | 9 | 1237 | |||
| BC017630.1 | - | 9 | 1180 | 4507289 | ||
| BC035548.1 | - | 9 | 1202 | 4221284 | ||

Rnf44
Genomic Map