siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035122.2 | NP_001030294.1 | 10 | 1178 | ||
| NM_027307.4 | NP_081583.3 | 11 | 1314 | |||
| GenBank mRNA | AK009072.1 | - | 9 | 985 | ||
| AK133865.1 | - | 10 | 1180 | |||
| AK167646.1 | - | 10 | 1155 | |||
| BC011152.1 | - | 10 | 1139 | 4188434 | ||
| BC098486.1 | - | 11 | 1310 | 30024953 | ||
| BU152451.1 | - | 2 | 56 | |||
| CN529028.1 | - | 2 | 116 | |||

Golm1
Genomic Map