siRNA Details
| Gene Symbol: Akr1c18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134066.2 | NP_598827.1 | 8, 9 | 956 | ||
| GenBank mRNA | AB059565.1 | - | 8, 9 | 956 | ||
| AK153898.1 | - | 8, 9 | 974 | |||
| BC034259.1 | - | 7, 8 | 875 | 4238589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akr1c18| Gene Symbol: Akr1c18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134066.2 | NP_598827.1 | 8, 9 | 956 | ||
| GenBank mRNA | AB059565.1 | - | 8, 9 | 956 | ||
| AK153898.1 | - | 8, 9 | 974 | |||
| BC034259.1 | - | 7, 8 | 875 | 4238589 | ||