siRNA Details
| Gene Symbol: AW209491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104646.1 | NP_001098116.1 | 2 | 866 | ||
| NM_134067.4 | NP_598828.3 | 2 | 933 | |||
| GenBank mRNA | AK030506.1 | - | 915 | |||
| AK050507.1 | - | 2 | 915 | |||
| AK077649.1 | - | 2 | 915 | |||
| AK167401.1 | - | 2 | 934 | |||
| AK169400.1 | - | 2 | 866 | |||
| BC016098.1 | - | 1 | 830 | 4910571 | ||
| BC034309.1 | - | 1 | 689 | 5362794 | ||

AW209491
Genomic Map