siRNA Details
Gene Symbol: E330034G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033214.2 | NP_001028386.1 | 6 | 937 | ||
GenBank mRNA | AK087874.1 | - | 6 | 737 | ||
AK136036.1 | - | 6 | 938 | |||
BC139082.1 | - | 6 | 908 | 8862104 | ||
BC139083.1 | - | 6 | 908 | 8862105 |
