siRNA Details
| Gene Symbol: Fam149b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024512.2 | NP_001019683.1 | 2 | 250 | ||
| NM_001177633.1 | NP_001171104.1 | 2 | 250 | |||
| NM_172379.3 | NP_759011.2 | 2 | 250 | |||
| GenBank mRNA | AK031973.1 | - | 2 | 253 | ||
| AK037808.1 | - | 2 | 138 | |||
| AK077495.1 | - | 2 | 153 | |||
| AK129252.1 | - | 2 | 173 | |||
| BB622169.1 | - | 2 | 252 | |||
| BC025183.1 | - | 2 | 146 | 5011464 | ||
| BC057907.1 | - | 2 | 152 | 5064176 | ||
| BC058650.1 | - | 2 | 93 | |||
| BC059221.1 | - | 2 | 93 | |||
| BC069846.1 | - | 2 | 148 | 4196478 | ||
| BC080849.1 | - | 2 | 190 | 5717789 | ||

Fam149b
Genomic Map