siRNA Details
Gene Symbol: Kctd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111028.1 | NP_001104498.1 | 11 | 1283 | ||
NM_001285933.1 | NP_001272862.1 | 11 | 1410 | |||
NM_134073.2 | NP_598834.2 | 10 | 1133 | |||
GenBank mRNA | AK158922.1 | - | 10 | 1135 | ||
BC011302.1 | - | 11 | 1224 | |||
CK618608.1 | - | 4 | 428 |
