siRNA Details
| Gene Symbol: Exoc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207214.3 | NP_997097.1 | 17 | 2359 | ||
| GenBank mRNA | AK134562.1 | - | 17 | 1872 | ||
| AK150297.1 | - | 6 | 626 | |||
| AK152307.1 | - | 17 | 2088 | |||
| AK157473.1 | - | 17 | 2054 | |||
| AK164067.1 | - | 17 | 2081 | |||
| AK167145.1 | - | 17 | 2047 | |||
| AK170148.1 | - | 17 | 2053 | |||
| AK171012.1 | - | 17 | 2077 | |||
| BC049967.1 | - | 17 | 2016 | 3672313 | ||

Exoc5
Genomic Map